Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ AKAP10 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA556007
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA556007 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA556007 Supplier Invitrogen™ Supplier No. PA556007
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: SAQLFMTHSE GIDLNNRTNS TQNHLLLSQE CDSAHSLRLE MARAGTHQVS METQESSSTL TVASRNSPAS PLKELSGKLM KSIEQDAVNT FTKYISPDAA KPIPITEAMR NDIIARICGE DGQVDPNCFV Highest antigen sequence identity to the following orthologs: Mouse - 75%, Rat - 80%.

The A-kinase anchor proteins are a group of structurally diverse proteins, which have the common function of binding to the regulatory subunit of protein kinase A and confining the holoenzyme to discrete locations within the cell. This gene encodes a member of the AKAP family. The encoded protein interacts with both the type I and type II regulatory subunits of PKA; therefore, it is a dual-specific AKAP. This protein is highly enriched in mitochondria. It contains RGS domains, in addition to a PKA-RII subunit-binding domain. The mitochondrial localization and the presence of RGS domains may have important implications for the function of this protein in PKA and G protein signal transduction.
TRUSTED_SUSTAINABILITY

Specifications

Antigen AKAP10
Applications Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry
Classification Polyclonal
Concentration 0.2 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene AKAP10
Gene Accession No. O43572
Gene Alias 1500031L16Rik; A kinase (PRKA) anchor protein 10; A kinase anchor protein 10; A kinase anchor protein 10, mitochondrial; Akap10; AKAP-10; A-kinase anchor protein 10, mitochondrial; A-kinase anchoring protein 10; B130049N18Rik; D-akap2; D-AKAP-2; Dual specific A kinase anchoring protein 2; dual specificity A kinase-anchoring protein 2; MGC9414; mitochondrial A kinase PPKA anchor protein 10; PRKA10; protein kinase A anchoring protein 10; protein kinase A-anchoring protein 10
Gene Symbols AKAP10
Host Species Rabbit
Immunogen Recombinant protein corresponding to Human AKAP10. Recombinant protein control fragment (Product #RP-102397).
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 11216
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.