Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ AKAP10 Polyclonal Antibody
Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA536131
Description
This antibody detects endogenous protein at a molecular weight of 74 kDa. Purity is >95% by SDS-PAGE.
The A-kinase anchor proteins are a group of structurally diverse proteins, which have the common function of binding to the regulatory subunit of protein kinase A and confining the holoenzyme to discrete locations within the cell. This gene encodes a member of the AKAP family. The encoded protein interacts with both the type I and type II regulatory subunits of PKA; therefore, it is a dual-specific AKAP. This protein is highly enriched in mitochondria. It contains RGS domains, in addition to a PKA-RII subunit-binding domain. The mitochondrial localization and the presence of RGS domains may have important implications for the function of this protein in PKA and G protein signal transduction.
Specifications
AKAP10 | |
Polyclonal | |
Unconjugated | |
AKAP10 | |
1500031L16Rik; A kinase (PRKA) anchor protein 10; A kinase anchor protein 10; A kinase anchor protein 10, mitochondrial; Akap10; AKAP-10; A-kinase anchor protein 10, mitochondrial; A-kinase anchoring protein 10; B130049N18Rik; D-akap2; D-AKAP-2; Dual specific A kinase anchoring protein 2; dual specificity A kinase-anchoring protein 2; MGC9414; mitochondrial A kinase PPKA anchor protein 10; PRKA10; protein kinase A anchoring protein 10; protein kinase A-anchoring protein 10 | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
11216 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Immunohistochemistry (Paraffin), Western Blot | |
1 mg/mL | |
PBS with 50% glycerol and 0.02% sodium azide; pH 7.2 | |
O43572 | |
AKAP10 | |
Synthetic peptide corresponding to amino acids 1-50 of human AKAP 10. | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Safety and Handling
WARNING: Cancer - www.P65Warnings.ca.gov
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction