Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ AKAP11 Polyclonal Antibody

Catalog No. PIPA5122256
Change view
Click to view available options
Quantity:
20 μL
200 μL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA5122256 20 μL
PIPA5118922 200 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. PIPA5122256 Supplier Invitrogen™ Supplier No. PA5122256
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

AKAP11 Polyclonal Antibody for Western Blot, ICC/IF, IHC (P)

The A-kinase anchor proteins (AKAPs) are a group of structurally diverse proteins, which have the common function of binding to the regulatory subunit of protein kinase A (PKA) and confining the holoenzyme to discrete locations within the cell. This gene encodes a member of the AKAP family. The encoded protein is expressed at high levels throughout spermatogenesis and in mature sperm. It binds the RI and RII subunits of PKA in testis. It may serve a function in cell cycle control of both somatic cells and germ cells in addition to its putative role in spermatogenesis and sperm function. [provided by RefSeq, Jul 2008].
TRUSTED_SUSTAINABILITY

Specifications

Antigen AKAP11
Applications Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry
Classification Polyclonal
Concentration 0.5 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.05% ProClin 300; pH 7.4
Gene AKAP11
Gene Accession No. Q9UKA4
Gene Alias A kinase (PRKA) anchor protein 11; a kinase anchor protein 220 kDa; AKAP 220; AKAP11; AKAP-11; AKAP220; A-kinase anchor protein 11; A-kinase anchor protein 220 kDa; A-kinase anchoring protein 11; A-kinase anchoring protein, 220kDa; KIAA0629; PPP1R44; PRKA11; protein kinase A anchoring protein 11; protein kinase A-anchoring protein 11; protein phosphatase 1, regulatory subunit 44
Gene Symbols AKAP11
Host Species Rabbit
Immunogen Recombinant protein A Kinase Anchor Protein 11. The antigen corresponds to amino acid range 1650-1901 of the target protein.
Purification Method Antigen affinity chromatography, Protein A
Quantity 20 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 11215
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.