Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ AKAP11 Polyclonal Antibody
Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA558652
Description
Immunogen sequence: EHSGKKVQFA EALATHILSL ATEMAASHLD NKIIQEPKVK NPCLNVQSQR SVSPTFLNPS DENLKTLCNF AGDLAAEVIT EAEKIAKVRN CM Highest antigen sequence identity to the following orthologs: Mouse - 67%, Rat - 64%.
The A-kinase anchor proteins (AKAPs) are a group of structurally diverse proteins, which have the common function of binding to the regulatory subunit of protein kinase A (PKA) and confining the holoenzyme to discrete locations within the cell. This gene encodes a member of the AKAP family. The encoded protein is expressed at high levels throughout spermatogenesis and in mature sperm. It binds the RI and RII subunits of PKA in testis. It may serve a function in cell cycle control of both somatic cells and germ cells in addition to its putative role in spermatogenesis and sperm function. [provided by RefSeq, Jul 2008].
Specifications
AKAP11 | |
Polyclonal | |
Unconjugated | |
AKAP11 | |
A kinase (PRKA) anchor protein 11; a kinase anchor protein 220 kDa; AKAP 220; AKAP11; AKAP-11; AKAP220; A-kinase anchor protein 11; A-kinase anchor protein 220 kDa; A-kinase anchoring protein 11; A-kinase anchoring protein, 220kDa; KIAA0629; PPP1R44; PRKA11; protein kinase A anchoring protein 11; protein kinase A-anchoring protein 11; protein phosphatase 1, regulatory subunit 44 | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
11215 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Immunohistochemistry (Paraffin) | |
0.4 mg/mL | |
PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
Q9UKA4 | |
AKAP11 | |
Recombinant protein corresponding to Human AKAP11. Recombinant protein control fragment (Product #RP-97912). | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction