Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
AKAP12 Antibody, Novus Biologicals™

Rabbit Polyclonal Antibody has been used in 1 publication
$382.00 - $610.00
Specifications
Antigen | AKAP12 |
---|---|
Applications | Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin) |
Classification | Polyclonal |
Conjugate | Unconjugated |
Host Species | Rabbit |
Description
AKAP12 Polyclonal specifically detects AKAP12 in Human samples. It is validated for Western Blot, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin.Specifications
AKAP12 | |
Polyclonal | |
Rabbit | |
Signal Transduction | |
PBS (pH 7.2) and 40% Glycerol with 0.02% Sodium Azide | |
A kinase (PRKA) anchor protein (gravin) 12, A kinase (PRKA) anchor protein 12, AKAP 250, AKAP250250kDa, DKFZp686M0430, DKFZp686O0331, FLJ20945, FLJ97621, Gravin, kinase scaffold protein gravin, Myasthenia gravis autoantigen, myasthenia gravis autoantigen gravin | |
AKAP12 | |
IgG | |
Affinity Purified | |
Specificity of antibody verified on a Protein Array containing target protein plus 383 other non-specific proteins. |
Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin) | |
Unconjugated | |
RUO | |
Human | |
Q02952 | |
9590 | |
This antibody was developed against Recombinant Protein corresponding to amino acids:QRAEAERPEEQAEASGLKKETDVVLKVDAQEAKTEPFTQGKVVGQTTPESFEKAPQVTESIESSELVTTCQAETLAGVKSQEMVMEQAIPPDSVETPTDSETDGST | |
Primary | |
Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. |
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title