Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

AKAP12/Gravin Rabbit anti-Human, Polyclonal, Bioss
SDP

Catalog No. 501983142
Encompass_Preferred
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-198-3142 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 50-198-3142 Supplier Bioss Supplier No. BS1381R
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

The A-kinase anchor proteins (AKAPs) are a group of structurally diverse proteins, which have the common function of binding to the regulatory subunit of protein kinase A (PKA) and confining the holoenzyme to discrete locations within the cell. This gene encodes a member of the AKAP family. The encoded protein is expressed in endothelial cells, cultured fibroblasts, and osteosarcoma cells. It associates with protein kinases A and C and phosphatase, and serves as a scaffold protein in signal transduction. This protein and RII PKA colocalize at the cell periphery. This protein is a cell growth-related protein. Antibodies to this protein can be produced by patients with myasthenia gravis. Alternative splicing of this gene results in two transcript variants encoding different isoforms.

Specifications

Antigen AKAP12/Gravin
Applications Immunofluorescence, Immunohistochemistry (Paraffin), Western Blot
Classification Polyclonal
Concentration 1 μg/mL
Conjugate Unconjugated
Formulation PBS with 1% BSA, 50% glycerol and 0.09% sodium azide; pH
Gene AKAP12
Gene Accession No. Q02952
Gene Alias A kinase (PRKA) anchor protein (gravin) 12; A kinase (PRKA) anchor protein 12; AI317366; AKAP 250; Akap12; AKAP-12; AKAP12A; AKAP12B; AKAP12G; AKAP250; A-kinase anchor protein 12; A-kinase anchor protein 250 kDa; A-kinase anchor protein, 250kDa; A-kinase anchoring protein 12; Gag12; gercelin; Germ cell lineage protein gercelin; Gravin; kinase scaffold protein gravin; Myasthenia gravis autoantigen; myasthenia gravis autoantigen gravin; Srcs5; Src-suppressed C kinase substrate; Ssecks; SSeCKS, Gravin, AKAP250; testis specific gene A12; Tsga12; v-src suppressed transcript 5
Gene Symbols AKAP12
Host Species Rabbit
Immunogen KLH conjugated synthetic peptide derived from human of human AKAP12.
Purification Method Protein A
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 9590
Target Species Human
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.