Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

AKAP12 Sheep anti-Rat, Polyclonal, Novus Biologicals™

Sheep Polyclonal Antibody

Manufacturer:  Novus Biologicals NBP266604

Catalog No. NBP266604


Add to cart

Description

Description

AKAP12 Polyclonal antibody specifically detects AKAP12 in Rat samples. It is validated for Western Blot, Immunohistochemistry, Immunoprecipitation.
Specifications

Specifications

AKAP12
Unconjugated
PBS with 0.08% Sodium Azide
A kinase (PRKA) anchor protein (gravin) 12, A kinase (PRKA) anchor protein 12, AKAP 250, AKAP250250kDa, DKFZp686M0430, DKFZp686O0331, FLJ20945, FLJ97621, Gravin, kinase scaffold protein gravin, Myasthenia gravis autoantigen, myasthenia gravis autoantigen gravin
Sheep
IgG
250μg
Store at -20°C. Avoid freeze-thaw cycles.
Polyclonal
Rat
Immunohistochemistry, Immunoprecipitation, Western Blot
Western Blot 1:100-1:2000, Immunohistochemistry 1:10 - 1:500, Immunoprecipitation 1:10 - 1:500
Purified
AKAP12
Recombinant protein corresponding to amino acids 620-1596 of the SSeCKS protein.
Ammonium sulfate precipitation
Signal Transduction
Primary
9590
SDS
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit