Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
AKAP13 Antibody, HRP, Novus Biologicals™

Rabbit Polyclonal Antibody
Supplier: Novus Biologicals NB10068213H
Description
AKAP13 Polyclonal antibody specifically detects AKAP13 in Human samples. It is validated for Western BlotSpecifications
AKAP13 | |
Polyclonal | |
Western Blot | |
A kinase (PRKA) anchor protein 13, AKAP-LbcAKAP-13, A-kinase anchor protein 13, A-kinase anchoring protein, ARHGEF13, Breast cancer nuclear receptor-binding auxiliary protein, BRXNon-oncogenic Rho GTPase-specific GTP exchange factor, c-lbc, FLJ11952, FLJ43341, Guanine nucleotide exchange factor Lbc, HA-3, HT31, Human thyroid-anchoring protein 31, LBCLBC oncogene, Lymphoid blast crisis oncogeneProtein kinase A-anchoring protein 13, p47, PROTO-LB, PROTO-LBC | |
The immunogen recognized by this antibody maps to a region between residue 1850 and 1900 of human A kinase (PRKA) anchor protein 13 using the numbering given in entry NP_009131.2 (GeneID 11214). | |
0.1 mL | |
Breast Cancer | |
11214 | |
Store at 4C in the dark. | |
IgG |
Western Blot | |
HRP | |
PBS | |
Rabbit | |
Immunogen affinity purified | |
RUO | |
Primary | |
Human | |
Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction