Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ AKAP13 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA5101150
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA5101150 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA5101150 Supplier Invitrogen™ Supplier No. PA5101150
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Antibody detects endogenous levels of total AKAP13.

The A-kinase anchor proteins (AKAPs) are a group of structurally diverse proteins which have the common function of binding to the regulatory subunit of protein kinase A (PKA) and confining the holoenzyme to discrete locations within the cell. This gene encodes a member of the AKAP family. Alternative splicing of this gene results in multiple transcript variants encoding different isoforms containing c-terminal dbl oncogene homology (DH) and pleckstrin homology (PH) domains. The DH domain is associated with guanine nucleotide exchange activation for the Rho/Rac family of small GTP binding proteins, resulting in the conversion of the inactive GTPase to the active form capable of transducing signals. The PH domain has multiple functions. Therefore, these isoforms function as scaffolding proteins to coordinate a Rho signaling pathway, function as protein kinase A-anchoring proteins and, in addition, enhance ligand-dependent activity of estrogen receptors alpha and beta.
TRUSTED_SUSTAINABILITY

Specifications

Antigen AKAP13
Applications Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry
Classification Polyclonal
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.02% sodium azide; pH 7.4
Gene AKAP13
Gene Accession No. E9Q394, Q12802
Gene Alias 1700026G02Rik; 5730522G15Rik; 5830460E08Rik; A kinase (PRKA) anchor protein 13; AKAP13; AKAP-13; AKAP-Lbc; A-kinase anchor protein 13; A-kinase anchoring protein; A-kinase anchoring protein 13; ARHGEF13; Breast cancer nuclear receptor-binding auxiliary protein; Brx; c-lbc; guanine nucleotide exchange factor Lbc; HA-3; Ht31; Human thyroid-anchoring protein 31; LBC; LBC oncogene; lymphoid blast crisis oncogene; Non-oncogenic Rho GTPase-specific GTP exchange factor; p47; PRKA13; Protein kinase A-anchoring protein 13; PROTO-LB; PROTO-LBC; type II cAMP-dependent protein kinase anchoring protein Ht31
Gene Symbols AKAP13
Host Species Rabbit
Immunogen A synthesized peptide derived from human AKAP13(Accession Q12802), corresponding to amino acid residues T718-K768.
Purification Method Affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 11214, 75547
Target Species Human, Mouse
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.