Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ AKAP13 Polyclonal Antibody

Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA554078
Description
Immunogen sequence: ARELKEQLHQ KDQKILLLLE EKEMIFRDMA ECSTPLPEDC SPTHSPRVLF RSNTEEALKG GPLMKSAINE VEILQGLVSG NLGGTLGPTV SSPIEQDVVG PVSLPR Highest antigen sequence identity to the following orthologs: Mouse - 87%, Rat - 87%.
The A-kinase anchor proteins (AKAPs) are a group of structurally diverse proteins which have the common function of binding to the regulatory subunit of protein kinase A (PKA) and confining the holoenzyme to discrete locations within the cell. This gene encodes a member of the AKAP family. Alternative splicing of this gene results in multiple transcript variants encoding different isoforms containing c-terminal dbl oncogene homology (DH) and pleckstrin homology (PH) domains. The DH domain is associated with guanine nucleotide exchange activation for the Rho/Rac family of small GTP binding proteins, resulting in the conversion of the inactive GTPase to the active form capable of transducing signals. The PH domain has multiple functions. Therefore, these isoforms function as scaffolding proteins to coordinate a Rho signaling pathway, function as protein kinase A-anchoring proteins and, in addition, enhance ligand-dependent activity of estrogen receptors alpha and beta.
Specifications
AKAP13 | |
Polyclonal | |
Unconjugated | |
AKAP13 | |
1700026G02Rik; 5730522G15Rik; 5830460E08Rik; A kinase (PRKA) anchor protein 13; AKAP13; AKAP-13; AKAP-Lbc; A-kinase anchor protein 13; A-kinase anchoring protein; A-kinase anchoring protein 13; ARHGEF13; Breast cancer nuclear receptor-binding auxiliary protein; Brx; c-lbc; guanine nucleotide exchange factor Lbc; HA-3; Ht31; Human thyroid-anchoring protein 31; LBC; LBC oncogene; lymphoid blast crisis oncogene; Non-oncogenic Rho GTPase-specific GTP exchange factor; p47; PRKA13; Protein kinase A-anchoring protein 13; PROTO-LB; PROTO-LBC; type II cAMP-dependent protein kinase anchoring protein Ht31 | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
11214 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Immunohistochemistry (Paraffin), Immunocytochemistry | |
0.3 mg/mL | |
PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
Q12802 | |
AKAP13 | |
Recombinant protein corresponding to Human AKAP13. Recombinant protein control fragment (Product #RP-102442). | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction