Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
AKAP13 Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Supplier: Thermo Scientific PA538670
Description
The type II cAMP-protein kinase (PKA) is a multifunctional kinase with a broad range of substrates. Specificity of PKA signaling is thought to be mediated by the compartmentalization of the kinase to specific sites within the cell. To maintain this specific localization, the R subunit (RII) of PKA interacts with specific RII-anchoring proteins. The family of RII-anchoring proteins has been designated A-kinase anchoring proteins (AKAP). AKAP 13, also known as Brx (breast cancer nuclear receptor-binding auxiliary protein), Lbc (lymphoid blast crisis oncogene), HA-3 or Ht31 (human thyroid-anchoring protein 31), functions as a cAMP-dependent scaffold anchor for PKA and also has Rho-GEF activity. It is known to regulate TLR2 signaling, NFkB activation, protein kinase D activation and participate in Actin stress fiber formation. Seven isoforms exist for AKAP 13 and, depending on the isoform, it localizes to the cytoplasm, nucleus or cell membrane.
Specifications
| AKAP13 | |
| Polyclonal | |
| Unconjugated | |
| AKAP13 | |
| A-kinase anchor protein 13, AKAP-13, AKAP-Lbc, Breast cancer nuclear receptor-binding auxiliary protein, Guanine nucleotide exchange factor Lbc, Human thyroid-anchoring protein 31, Lymphoid blast crisis oncogene, LBC oncogene, Non-oncogenic Rho GTPase-specific GTP exchange factor, Protein kinase A-anchoring protein 13, PRKA13, p47, BRX, HT31, LBC, AKAP13 | |
| Rabbit | |
| Antigen affinity chromatography | |
| RUO | |
| 11214 | |
| -20°C | |
| Liquid |
| Immunohistochemistry (Paraffin), Western Blot | |
| 1 mg/mL | |
| Dulbecco′s PBS with 150mM NaCl, 50% glycerol and 0.02% sodium azide; pH 7.4 | |
| Q12802 | |
| AKAP13 | |
| A synthetic peptide derived from the internal region of human AKAP13 | |
| 100 μg | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Safety and Handling
WARNING: Cancer - www.P65Warnings.ca.gov
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction