Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ AKAP14 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA599280
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA599280 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA599280 Supplier Invitrogen™ Supplier No. PA599280
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Antibody detects endogenous levels of total AKAP14.

The A-kinase anchor proteins (AKAPs) are a group of structurally diverse proteins, which have the common function of binding to the regulatory subunit of protein kinase A (PKA) and confining the holoenzyme to discrete locations within the cell. This gene encodes a member of the AKAP family. The protein anchors PKA in ciliary axonemes and, in this way, may play a role in regulating ciliary beat frequency. Alternate transcriptional splice variants, encoding different isoforms, have been characterized.
TRUSTED_SUSTAINABILITY

Specifications

Antigen AKAP14
Applications Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry
Classification Polyclonal
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.02% sodium azide; pH 7.4
Gene AKAP14
Gene Accession No. Q86UN6
Gene Alias A kinase (PRKA) anchor protein 14; AKAP 28; AKAP14; AKAP-14; AKAP28; A-kinase anchor protein 14; A-kinase anchor protein 28 kDa; A-kinase anchoring protein 14; A-kinase anchoring protein 28; PRKA14; Protein kinase A-anchoring protein 14; TAKAP-1.2; Takap80; Testis-specific A-kinase-anchoring protein TAKAP-80; Testis-specific A-kinase-anchoring-protein
Gene Symbols AKAP14
Host Species Rabbit
Immunogen A synthesized peptide derived from human AKAP14(Accession Q86UN6), corresponding to amino acid residues M1-D34.
Purification Method Affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 158798
Target Species Human
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.