Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ AKAP17A Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA560291
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA560291 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA560291 Supplier Invitrogen™ Supplier No. PA560291
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: GEVENKSLVK SFLACLDGKT IKLSGFSDIL KVRAAEFKID FPTRHDWDSF FRDAKDMNET LPGERPDTIH LEGLPCKWFA LKESGSEKPS EDVLVKVF Highest antigen sequence identity to the following orthologs: Mouse - 41%, Rat - 41%.

DXYS155E is a gene found in the pseudoautosomal region of the distal short arms of the X and Y chromosomes, and appears to be ubiquitously expressed.
TRUSTED_SUSTAINABILITY

Specifications

Antigen AKAP17A
Applications Immunohistochemistry (Paraffin)
Classification Polyclonal
Concentration 0.1 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene AKAP17A
Gene Accession No. Q02040
Gene Alias 721P; A kinase (PRKA) anchor protein 17A; AKAP17A; AKAP-17A; A-kinase anchor protein 17A; A-kinase anchoring protein 17A; B-lymphocyte antigen; B-lymphocyte surface antigen; CCDC133; CXYorf3; DXYS155E; PRKA17A; Protein kinase A-anchoring protein 17A; Protein XE7; pseudoautosomal gene XE7; RP13-297E16.1; SFRS17A; Splicing factor, arginine/serine-rich 17A; XE7; XE7Y
Gene Symbols AKAP17A
Host Species Rabbit
Immunogen Recombinant protein corresponding to Human AKAP17A. Recombinant protein control fragment (Product #RP-100086).
Purification Method Antigen Affinity Chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 8227
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.