Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

AKAP17A Polyclonal Antibody, Invitrogen™

Catalog No. PIPA540844
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA540844 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA540844 Supplier Thermo Scientific Supplier No. PA540844

Rabbit Polyclonal Antibody

Peptide sequence: NWEVMERLKG MVQNHQFSTL RISKSTMDFI RFEGEVENKS LVKSFLACLD Sequence homology: Dog: 100%; Horse: 100%; Human: 100%; Rat: 93%

AKAP17A Polyclonal antibody specifically detects AKAP17A in Human samples. It is validated for Immunohistochemistry, Western Blot.
TRUSTED_SUSTAINABILITY

Specifications

Antigen AKAP17A
Applications Immunohistochemistry, Western Blot
Classification Polyclonal
Concentration 0.5 mg/mL
Conjugate Unconjugated
Formulation PBS with 2% sucrose and 0.09% sodium azide
Gene AKAP17A
Gene Accession No. Q02040
Gene Alias 721P; A kinase (PRKA) anchor protein 17A; AKAP17A; AKAP-17A; A-kinase anchor protein 17A; A-kinase anchoring protein 17A; B-lymphocyte antigen; B-lymphocyte surface antigen; CCDC133; CXYorf3; DXYS155E; PRKA17A; Protein kinase A-anchoring protein 17A; Protein XE7; pseudoautosomal gene XE7; RP13-297E16.1; SFRS17A; Splicing factor, arginine/serine-rich 17A; XE7; XE7Y
Gene Symbols AKAP17A
Host Species Rabbit
Immunogen Synthetic peptide directed towards the N-terminal of human DXYS155E
Purification Method Affinity Chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 8227
Target Species Human
Content And Storage -20° C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.