Learn More
Invitrogen™ AKAP17A Polyclonal Antibody

Description
Recommended positive controls: 293T, HeLaS3, Raji. Predicted reactivity: Rhesus Monkey (87%). Store product as a concentrated solution. Centrifuge briefly prior to opening the vial.
Specifications
Specifications
| Antigen | AKAP17A |
| Applications | Immunohistochemistry (Paraffin), Western Blot |
| Classification | Polyclonal |
| Concentration | 1 mg/mL |
| Conjugate | Unconjugated |
| Formulation | 0.1M tris glycine with 10% glycerol and 0.01% thimerosal; pH 7 |
| Gene | AKAP17A |
| Gene Accession No. | Q02040 |
| Gene Alias | 721P; A kinase (PRKA) anchor protein 17A; AKAP17A; AKAP-17A; A-kinase anchor protein 17A; A-kinase anchoring protein 17A; B-lymphocyte antigen; B-lymphocyte surface antigen; CCDC133; CXYorf3; DXYS155E; PRKA17A; Protein kinase A-anchoring protein 17A; Protein XE7; pseudoautosomal gene XE7; RP13-297E16.1; SFRS17A; Splicing factor, arginine/serine-rich 17A; XE7; XE7Y |
| Gene Symbols | AKAP17A |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.