Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
AKAP2 Antibody, Janelia Fluor™ 549, Novus Biologicals™

Rabbit Polyclonal Antibody
Supplier: Novus Biologicals NB10061604JF549
Description
AKAP2 Polyclonal antibody specifically detects AKAP2 in Human, Sheep samples. It is validated for Western BlotSpecifications
AKAP2 | |
Polyclonal | |
Western Blot | |
A kinase (PRKA) anchor protein 2, AKAP-2, FLJ53197, protein kinase A2 | |
The immunogen recognized by this antibody maps to a region between residue 525 and 575 of human A kinase (PRKA) anchor protein 2 using the numbering given in entry Q9Y2D5.2 (GeneID 11217). | |
0.1 mL | |
Protein Kinase | |
445815 | |
Store at 4C in the dark. | |
IgG |
Western Blot | |
Janelia Fluor 549 | |
50mM Sodium Borate | |
Rabbit | |
Immunogen affinity purified | |
RUO | |
Primary | |
Human, Sheep | |
Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction