Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

AKAP2 Rabbit anti-Human, Polyclonal, Bioss
SDP

Catalog No. 501988764
Encompass_Preferred
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-198-8764 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 50-198-8764 Supplier Bioss Supplier No. BS9028R
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

The protein encoded by this gene binds to the regulatory subunit of protein kinase A and is found associated with the actin cytoskeleton. The encoded protein mediates signals carried by cAMP and may be involved in creating polarity in certain signaling processes. Three transcript variants encoding different isoforms have been found for this gene. [provided by RefSeq, Mar 2011].

Specifications

Antigen AKAP2
Applications Immunofluorescence, Immunohistochemistry (Paraffin), Western Blot
Classification Polyclonal
Concentration 1 μg/mL
Conjugate Unconjugated
Formulation PBS with 1% BSA, 50% glycerol and 0.09% sodium azide; pH
Gene AKAP2
Gene Accession No. Q9Y2D5
Gene Alias A kinase (PRKA) anchor protein 2; AKAP2; AKAP-2; AKAPKL; A-kinase anchor protein 2; A-kinase anchoring protein 2; KL2B; MISP2; PALM2-AKAP2; PRKA2; protein kinase A anchoring protein 2; protein kinase A2
Gene Symbols AKAP2
Host Species Rabbit
Immunogen KLH conjugated synthetic peptide derived from human AKAP2.
Purification Method Protein A
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 11217
Target Species Human
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.