Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ AKAP3 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA5109376
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA5109376 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA5109376 Supplier Invitrogen™ Supplier No. PA5109376
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: MSEKVDWLQS QNGVCKVDVY SPGDNQAQDW KMDTSTDPVR VLSWLRRDLE KSTAEFQDVR FKPGESFGGE TSNSGDPHKG FSVDYYNTTT KGTPERLHFE MTHKEIPCQG PRAQLGNGSS VDEVSFYANR LTNLVIAMAR KEINEKIDGS ENKCVYQSLY MGNEPTPTKS LSKIASELVN ETVSACSRNA APDKAPGSGD RVSGSSQSPP NLKYKSTLKI KESTKERQGP DDKPPSKKSF FYKEVFESRN GDYAREGGRF FPRERKRFRG QERPDDFTAS VSEGIMTYAN SVVSDMMVSI.

The type II cAMP-dependent protein kinase (PKA) is a multifunctional kinase with a broad range of substrates. Specificity of PKA signaling is mediated by the compartmentalization of the kinase to specific sites within the cell. To maintain this specific localization, the R subunit (RII) of PKA interacts with specific RII-anchoring proteins, designated A-kinase anchoring proteins (AKAP). AKAP 3, also known as AKAP 110, FSP95, PRKA3 and SOB1, binds both PKA and PDE4A and functions as a scaffolding protein in spermatozoa to regulate local cAMP concentrations and modulate sperm functions. Expression of AKAP 3 in normal tissues is restricted to the testis, where bicarbonate stimulates tyrosine phosphorylation of AKAP 3, thereby increasing its recruitment of PKA. AKAP-3 also exhibits high expression in patients with epithelial ovarian cancer (EOC). It demonstrates tumor-restricted expression and appears to be associated with worse overall survival, which make AKAP 3 a potential target for antigen-specific immunotherapy in EOC.
TRUSTED_SUSTAINABILITY

Specifications

Antigen AKAP3
Applications ELISA, Western Blot
Classification Polyclonal
Concentration 3.73 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.01% thimerosal; pH 7.3
Gene AKAP3
Gene Accession No. O88987
Gene Alias A kinase (PRKA) anchor protein 3; AKAP 110; AKAP110; Akap3; AKAP-3; A-kinase anchor protein 110 kDa; A-kinase anchor protein 3; A-kinase anchor protein, 110kDa; A-kinase anchoring protein 3; Cancer/testis antigen 82; CT82; epididymis luminal protein 159; Fibrous Sheath Protein of 95 kDa; fibrousheathin 1; Fibrousheathin I; fibrousheathin-1; FSP95; HEL159; PRKA3; protein kinase A binding protein AKAP 110; Protein kinase A-anchoring protein 3; SOB1; Sperm oocyte-binding protein; sperm oocyte-binding protein 1
Gene Symbols AKAP3
Host Species Rabbit
Immunogen Recombinant fusion protein containing a sequence corresponding to amino acids 1-300 of human AKAP3 (NP_006413.3).
Purification Method Affinity Chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 11642, 312720
Target Species Mouse, Rat
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.