Learn More
Invitrogen™ AKAP4 Polyclonal Antibody

Description
Immunogen sequence: QSPSAPPAKP PSTQRAVISP DGECSIDDLS FYVNRLSSLV IQMAHKEIKE KLEGKSKCLH HSICPSPGNK ERISPRTPAS KIASEMAYEA VELTAAEMRG TGEESREGGQ KSFLYSELSN KSKSGDKQMS QRESKEFADS ISKGLMVYAN QV.
Specifications
Specifications
| Antigen | AKAP4 |
| Applications | ELISA, Western Blot, Immunocytochemistry |
| Classification | Polyclonal |
| Concentration | 2.14 mg/mL |
| Conjugate | Unconjugated |
| Formulation | PBS with 50% glycerol and 0.05% ProClin 300; pH 7.3 |
| Gene | AKAP4 |
| Gene Accession No. | O35774, Q5JQC9, Q60662 |
| Gene Alias | 75 kDa fibrous sheath protein; A kinase (PRKA) anchor protein 4; AKAP 82; AKAP4; AKAP-4; akap4 {ECO:0000312; AKAP82; A-kinase anchor protein 4; A-kinase anchor protein 82 kDa; A-kinase anchoring protein 4; cancer/testis antigen 99; CT99; Fsc1; hAKAP82; HI; major sperm fibrous sheath protein; mAKAP82; p82; PRKA4; protein kinase A anchoring protein 4; protein kinase A-anchoring protein 4; RGD:620828}; testis-specific gene HI |
| Gene Symbols | AKAP4 |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.