Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ AKAP4 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA5114834
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA5114834 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA5114834 Supplier Invitrogen™ Supplier No. PA5114834
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Antibody detects endogenous levels of total AKAP4.

The A-kinase anchor proteins (AKAPs) are a group of structurally diverse proteins, which have the common function of binding to the regulatory subunit of protein kinase A (PKA) and confining the holoenzyme to discrete locations within the cell. AKAP4, also known as hAKAP82, is a major sperm fibrous sheath protein that localizes to the entire length of the flagellum of human sperm. It has been suggested that AKAP4 plays multiple roles in sperm motility and regulation of signal transduction pathways. AKAP4 has also been postulated to be a potential biomarker for breast cancer.
TRUSTED_SUSTAINABILITY

Specifications

Antigen AKAP4
Applications Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry
Classification Polyclonal
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.02% sodium azide
Gene AKAP4
Gene Accession No. Q5JQC9
Gene Alias 75 kDa fibrous sheath protein; A kinase (PRKA) anchor protein 4; AKAP 82; AKAP4; AKAP-4; akap4 {ECO:0000312; AKAP82; A-kinase anchor protein 4; A-kinase anchor protein 82 kDa; A-kinase anchoring protein 4; cancer/testis antigen 99; CT99; Fsc1; hAKAP82; HI; major sperm fibrous sheath protein; mAKAP82; p82; PRKA4; protein kinase A anchoring protein 4; protein kinase A-anchoring protein 4; RGD:620828}; testis-specific gene HI
Gene Symbols AKAP4
Host Species Rabbit
Immunogen A synthesized peptide derived from human AKAP4(Accession Q5JQC9), corresponding to amino acid residues A128-E178.
Purification Method Affinity Chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 8852
Target Species Human
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.