Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

AKAP4 Rabbit anti-Human, Polyclonal, Proteintech
SDP

Catalog No. 501734654 Shop All Proteintech Group Inc Products
Encompass_Preferred
Change view
Click to view available options
Quantity:
150 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-173-4654 150 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 50-173-4654 Supplier Proteintech Group Inc Supplier No. 249861AP150UL
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

The A-kinase anchor proteins (AKAPs) are a group of structurally diverse proteins, which have the common function of binding to the regulatory subunit of protein kinase A (PKA) and confining the holoenzyme to discrete locations within the cell. AKAP4, also known as hAKAP82, is a major sperm fibrous sheath protein that localizes to the entire length of the flagellum of human sperm. It has been suggested that AKAP4 plays multiple roles in sperm motility and regulation of signal transduction pathways. AKAP4 has also been postulated to be a potential biomarker for breast cancer.

Specifications

Antigen AKAP4
Applications Immunohistochemistry (Paraffin), Western Blot
Classification Polyclonal
Concentration 0.31 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.1% sodium azide; pH 7.3
Gene AKAP4
Gene Accession No. Q5JQC9
Gene Alias A kinase anchor protein 4, A kinase anchor protein 82 kDa, AKAP 4, AKAP 82, AKAP4, AKAP82, CT99, FSC1, hAKAP82, HI, p82, PRKA4
Gene Symbols AKAP4
Host Species Rabbit
Immunogen AKAP4 Fusion Protein Ag21762
Purification Method Antigen Affinity Chromatography
Quantity 150 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 8852
Target Species Human
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.