Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
AKAP6 Antibody, Novus Biologicals™

Rabbit Polyclonal Antibody
$382.00 - $610.00
Specifications
Antigen | AKAP6 |
---|---|
Dilution | Immunohistochemistry, Immunohistochemistry-Paraffin 1:1000 - 1:2500 |
Applications | Immunohistochemistry, Immunohistochemistry (Paraffin) |
Classification | Polyclonal |
Conjugate | Unconjugated |
Description
AKAP6 Polyclonal specifically detects AKAP6 in Human samples. It is validated for Immunohistochemistry, Immunohistochemistry-Paraffin.Specifications
AKAP6 | |
Immunohistochemistry, Immunohistochemistry (Paraffin) | |
Unconjugated | |
RUO | |
PBS (pH 7.2) and 40% Glycerol with 0.02% Sodium Azide | |
A kinase (PRKA) anchor protein 6, ADAP6A-kinase anchor protein 6, AKAP100A-kinase anchor protein 100 kDa, AKAP-6, KIAA0311AKAP 100, mAKAPMGC165020, PRKA6ADAP100, protein kinase A anchoring protein 6, Protein kinase A-anchoring protein 6 | |
AKAP6 | |
IgG | |
Affinity Purified |
Immunohistochemistry, Immunohistochemistry-Paraffin 1:1000 - 1:2500 | |
Polyclonal | |
Rabbit | |
Human | |
Q13023 | |
9472 | |
This antibody was developed against a recombinant protein corresponding to amino acids: SHSATKSALIQKLMQDIQHQDNYEAIWEKIEGFVNKLDEFIQWLNEAMETTENWTPPKAEMDDLKLYLETHLSFKLNVDSHCALKEAVEEEGHQLLELIASHKAGLKDMLRMIASQWKELQRQIKRQHSWILRALDTIKA | |
Primary | |
Specificity of antibody verified on a Protein Array containing target protein plus 383 other non-specific proteins. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title