Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ AKAP6 Monoclonal Antibody (2E8)
GREENER_CHOICE

Catalog No. PIMA524570
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIMA524570 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIMA524570 Supplier Invitrogen™ Supplier No. MA524570
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

Peptide Sequence: RAEVGKEVNG LPQTSSGCAE NLEFTPSKLD SEKESSGKPG ESGMPEEHNA ASAKSKVQDL SLKANQPTDK AALHPSPKTL TCEENLLNLH EKRHRNMH.

The A-kinase anchor proteins (AKAPs) are a group of structurally diverse proteins, which have the common function of binding to the regulatory subunit of protein kinase A (PKA) and confining the holoenzyme to discrete locations within the cell. This gene encodes a member of the AKAP family. The encoded protein is highly expressed in various brain regions and cardiac and skeletal muscle. It is specifically localized to the sarcoplasmic reticulum and nuclear membrane, and is involved in anchoring PKA to the nuclear membrane or sarcoplasmic reticulum.
TRUSTED_SUSTAINABILITY

Specifications

Antigen AKAP6
Applications ELISA, Western Blot
Classification Monoclonal
Clone 2E8
Conjugate Unconjugated
Formulation PBS with no preservative; pH 7.4
Gene AKAP6
Gene Accession No. Q13023
Gene Alias A kinase (PRKA) anchor protein 6; ADAP100; ADAP6; AI482140; AKAP 100; AKAP100; AKAP6; AKAP-6; Akapalpha; Akapbeta; A-kinase anchor protein 100 kDa; A-kinase anchor protein 6; A-kinase anchoring protein 6; A-kinase anchoring protein alpha; A-kinase anchoring protein beta; anchor protein 6; KIAA0311; mAkap; PRKA; PRKA6; protein kinase A anchoring protein 6; protein kinase A-anchoring protein 6
Gene Symbols AKAP6
Host Species Mouse
Immunogen AKAP6 (NP_004265.3, 2221 a.a. ~ 2318 a.a) partial recombinant protein with GST tag. MW of the GST tag alone is 26 KDa.
Purification Method Protein A
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 9472
Target Species Human
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG2b κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.