Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ AKAP6 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA5103319
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA5103319 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA5103319 Supplier Invitrogen™ Supplier No. PA5103319
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Antibody detects endogenous levels of total AKAP6.

The A-kinase anchor proteins (AKAPs) are a group of structurally diverse proteins, which have the common function of binding to the regulatory subunit of protein kinase A (PKA) and confining the holoenzyme to discrete locations within the cell. This gene encodes a member of the AKAP family. The encoded protein is highly expressed in various brain regions and cardiac and skeletal muscle. It is specifically localized to the sarcoplasmic reticulum and nuclear membrane, and is involved in anchoring PKA to the nuclear membrane or sarcoplasmic reticulum.
TRUSTED_SUSTAINABILITY

Specifications

Antigen AKAP6
Applications Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry
Classification Polyclonal
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.02% sodium azide; pH 7.4
Gene AKAP6
Gene Accession No. Q13023
Gene Alias A kinase (PRKA) anchor protein 6; ADAP100; ADAP6; AI482140; AKAP 100; AKAP100; AKAP6; AKAP-6; Akapalpha; Akapbeta; A-kinase anchor protein 100 kDa; A-kinase anchor protein 6; A-kinase anchoring protein 6; A-kinase anchoring protein alpha; A-kinase anchoring protein beta; anchor protein 6; KIAA0311; mAkap; PRKA; PRKA6; protein kinase A anchoring protein 6; protein kinase A-anchoring protein 6
Gene Symbols AKAP6
Host Species Rabbit
Immunogen A synthesized peptide derived from human AKAP6(Accession Q13023), corresponding to amino acid residues P2232-L2282.
Purification Method Affinity Chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 238161, 9472
Target Species Human, Mouse
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.