Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ AKAP6 Polyclonal Antibody

Goat Polyclonal Antibody
Supplier: Invitrogen™ PA5142104
This item is not returnable.
View return policy
Description
This antibody is tested in Peptide ELISA: antibody detection limit dilution 1:16,000.
The A-kinase anchor proteins (AKAPs) are a group of structurally diverse proteins, which have the common function of binding to the regulatory subunit of protein kinase A (PKA) and confining the holoenzyme to discrete locations within the cell. This gene encodes a member of the AKAP family. The encoded protein is highly expressed in various brain regions and cardiac and skeletal muscle. It is specifically localized to the sarcoplasmic reticulum and nuclear membrane, and is involved in anchoring PKA to the nuclear membrane or sarcoplasmic reticulum.
Specifications
AKAP6 | |
Polyclonal | |
Unconjugated | |
AKAP6 | |
A kinase (PRKA) anchor protein 6; ADAP100; ADAP6; AI482140; AKAP 100; AKAP100; AKAP6; AKAP-6; Akapalpha; Akapbeta; A-kinase anchor protein 100 kDa; A-kinase anchor protein 6; A-kinase anchoring protein 6; A-kinase anchoring protein alpha; A-kinase anchoring protein beta; anchor protein 6; KIAA0311; mAkap; PRKA; PRKA6; protein kinase A anchoring protein 6; protein kinase A-anchoring protein 6 | |
Goat | |
Ammonium sulfate precipitation | |
RUO | |
238161 | |
-20°C, Avoid Freeze/Thaw Cycles | |
Liquid |
Western Blot | |
0.5 mg/mL | |
TBS with 0.5% BSA and 0.02% sodium azide; pH 7.3 | |
0 | |
AKAP6 | |
Peptide with sequence LTMSVTLSPLRSQ-C (aa 2-14). | |
100 μg | |
Primary | |
Mouse | |
Antibody | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction