Learn More
Invitrogen™ AKAP6 Polyclonal Antibody

Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA588760
Description
Immunogen sequence: MLTMSVTLSP LRSQDLDPMA TDASPMAINM TPTVEQGEGE EAMKDMDSDQ QYEKPPPLHT GADWKIVLHL PEIETWLRMT SERVRDLTYS VQQDSDSKHV DVHLVQLKDI CEDISDHVEQ IHALLETEFS LKLLSYSVNV IVDIHAVQLL WHQLRVSVLV LRERILQGLQ DANGNYTRQT DILQAFSEET KEGRLDSLTE VDDSGQLTIK CSQNYLSLDC GITAFELSDY SPSEDLLSGL GDMTSSQVKT KPFDSWSYSE MEKEFPELIR; Positive Samples: U-87MG, SH-SY5Y; Cellular Location: Nucleus membrane, Sarcoplasmic reticulum.
The A-kinase anchor proteins (AKAPs) are a group of structurally diverse proteins, which have the common function of binding to the regulatory subunit of protein kinase A (PKA) and confining the holoenzyme to discrete locations within the cell. This gene encodes a member of the AKAP family. The encoded protein is highly expressed in various brain regions and cardiac and skeletal muscle. It is specifically localized to the sarcoplasmic reticulum and nuclear membrane, and is involved in anchoring PKA to the nuclear membrane or sarcoplasmic reticulum.
Specifications
| AKAP6 | |
| Polyclonal | |
| Unconjugated | |
| AKAP6 | |
| A kinase (PRKA) anchor protein 6; ADAP100; ADAP6; AI482140; AKAP 100; AKAP100; AKAP6; AKAP-6; Akapalpha; Akapbeta; A-kinase anchor protein 100 kDa; A-kinase anchor protein 6; A-kinase anchoring protein 6; A-kinase anchoring protein alpha; A-kinase anchoring protein beta; anchor protein 6; KIAA0311; mAkap; PRKA; PRKA6; protein kinase A anchoring protein 6; protein kinase A-anchoring protein 6 | |
| Rabbit | |
| Affinity Chromatography | |
| RUO | |
| 9472 | |
| -20°C, Avoid Freeze/Thaw Cycles | |
| Liquid |
| ELISA, Western Blot | |
| 1.08 mg/mL | |
| PBS with 50% glycerol and 0.01% thimerosal; pH 7.3 | |
| Q13023 | |
| AKAP6 | |
| Recombinant fusion protein containing a sequence corresponding to amino acids 1-270 of human AKAP6 (NP_0042653). | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.