Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ AKAP8 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA5116744
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA5116744 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA5116744 Supplier Invitrogen™ Supplier No. PA5116744
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Positive Samples: BT-474, HL-60, SKOV3, HeLa Immunogen sequence: EGAPAPESSG EPAEDEGPTD TAEAGSDPQA EQLLEEQVPC GTAHEKGVPK ARSEAAEAGN GAETMAAEAE SAQTRVAPAP AAADAEVEQT DAESKDAVPT E.

The type II cAMP-protein kinase (PKA) is a multifunctional kinase with a broad range of substrates. Specificity of PKA signaling is thought to be mediated by the compartmentalization of the kinase to specific sites within the cell. To maintain this specific localization, the R subunit (RII) of PKA interacts with specific RII-anchoring proteins. The family of RII-anchoring proteins has been designated A-kinase anchoring proteins (AKAP). AKAP 95, also known as AKAP 8, is a nuclear matrix protein predominantly expressed in liver, heart, pancreas, kidney and skeletal muscle. During mitosis, AKAP 95 is recruited to the chromosomes and plays an essential role in mitotic progression. Characteristic of its family, AKAP 95 participates in PKA signaling through an interaction with the RII regulatory subunit. In addition, AKAP 95 forms a complex with HA95 and HDAC3 and is required for the deacetylation of Histone H3 in mitosis.
TRUSTED_SUSTAINABILITY

Specifications

Antigen AKAP8
Applications ELISA, Immunohistochemistry (Paraffin), Western Blot
Classification Polyclonal
Concentration 0.49 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.02% sodium azide; pH 7.3
Gene AKAP8
Gene Accession No. O43823, Q63014
Gene Alias 1200016A02Rik; A kinase (PRKA) anchor protein 8; A kinase anchor protein 8; AA673585; AKAP 8; AKAP 95; Akap8; AKAP-8; Akap95; AKAP-95; A-kinase anchor protein 8; A-kinase anchor protein 95 kDa; A-kinase anchor protein, 95kDa; A-kinase anchoring protein 8; AU015639; DKFZp586B1222
Gene Symbols AKAP8
Host Species Rabbit
Immunogen A synthetic peptide corresponding to a sequence within amino acids 592-692 of human AKAP8 (NP_005849.1).
Purification Method Affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 10270, 116633
Target Species Human, Rat
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.