Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

AKAP8L/HA95 Polyclonal Antibody, Bethyl Laboratories

Catalog No. 501571809
Encompass_Preferred
Change view
Click to view available options
Quantity:
10 μL
100 μL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-157-1809 100 μL
50-157-1810 10 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 50-157-1809 Supplier Bethyl Laboratories, Inc Supplier No. IHC00277
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

The recommended shelf life for this product is 1 year from date of receipt. Based on 100% sequence identity, this antibody is predicted to react with Gorilla, Chimpanzee, Northern white-cheeked gibbon, Naked mole rat and Chinese hamster

Could play a role in constitutive transport element (CTE)-mediated gene expression. Does not seem to be implicated in the binding of regulatory subunit II of PKA. May be involved in nuclear envelope breakdown and chromatin condensation. May regulate the initiation phase of DNA replication when associated with TMPO-beta.

Specifications

Antigen AKAP8L/HA95
Applications Immunofluorescence, Immunohistochemistry
Classification Polyclonal
Conjugate Unconjugated
Formulation TBS with 0.1% BSA and 0.09% sodium azide
Gene AKAP8L
Gene Accession No. Q9ULX6
Gene Alias HA95, HAP95, NAKAP, NAKAP95, A-kinase anchor protein 8-like, AKAP8-like protein, helicase A-binding protein 95 kDa, homologous to AKAP95 protein, neighbor of A kinase anchoring protein 95, neighbor of A-kinase anchoring protein 95, neighbor of A-kinase-anchoring protein 95, neighbor of AKAP
Gene Symbols AKAP8L
Host Species Rabbit
Immunogen Between 596 and 646
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 26993
Target Species Human
Content And Storage 4° C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.