Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ AKAP8L Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA560018
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA560018 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA560018 Supplier Invitrogen™ Supplier No. PA560018
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: FLQEYVTNKT KKTEELRKTV EDLDGLIQQI YRDQDLTQEI AMEHFVKKVE AAHCAACDLF IPMQFGIIQK HLKTMDHNRN RR Highest antigen sequence identity to the following orthologs: Mouse - 100%, Rat - 99%.

Could play a role in constitutive transport element (CTE)-mediated gene expression by association with DHX9. Increases CTE-dependent nuclear unspliced mRNA export (PubMed:10748171, PubMed:11402034). Proposed to target PRKACA to the nucleus but does not seem to be implicated in the binding of regulatory subunit II of PKA (PubMed:10761695, PubMed:11884601). May be involved in nuclear envelope breakdown and chromatin condensation. May be involved in anchoring nuclear membranes to chromatin in interphase and in releasing membranes from chromating at mitosis (PubMed:11034899). May regulate the initiation phase of DNA replication when associated with TMPO isoform Beta (PubMed:12538639). Required for cell cycle G2/M transition and histone deacetylation during mitosis. In mitotic cells recruits HDAC3 to the vicinity of chromatin leading to deacetylation and subsequent phosphorylation at 'Ser-10' of histone H3; in this function seems to act redundantly with AKAP8 (PubMed:16980585). May be involved in regulation of pre-mRNA splicing (PubMed:17594903). [UniProt].
TRUSTED_SUSTAINABILITY

Specifications

Antigen AKAP8L
Applications Immunohistochemistry (Paraffin)
Classification Polyclonal
Concentration 0.1 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene AKAP8L
Gene Accession No. Q9ULX6
Gene Alias A kinase (PRKA) anchor protein 8-like; AKAP8L; AKAP8-like protein; A-kinase anchor protein 8-like; A-kinase anchoring protein 8 like; HA95; HAP95; Helicase A-binding protein 95; helicase A-binding protein 95 kDa; homologous to AKAP95 protein; HRIHFB2018; LOC539700 protein; NAKAP; NAKAP95; neighbor of AKAP; Neighbor of AKAP95; neighbor of A-kinase anchoring protein 95; neighbor of A-kinase-anchoring protein 95; testis tissue sperm-binding protein Li 90mP
Gene Symbols AKAP8L
Host Species Rabbit
Immunogen Recombinant protein corresponding to Human AKAP8L. Recombinant protein control fragment (Product #RP-98866).
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 26993
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.