Learn More
AKAP9/CG-NAP Polyclonal Antibody, Bethyl Laboratories
Description
The recommended shelf life for this product is 1 year from date of receipt.
Specifications
Specifications
| Antigen | AKAP9/CG-NAP |
| Applications | Western Blot |
| Classification | Polyclonal |
| Conjugate | Unconjugated |
| Formulation | TBS with 0.1% BSA and 0.09% sodium azide |
| Gene | AKAP9 |
| Gene Accession No. | Q99996 |
| Gene Alias | AKAP9, CG-NAP, PRKA9, YOTIAO, AKAP350, HYPERION, KIAA0803, A kinase (PRKA) anchor protein (yotiao) 9, AKAP120-like protein, kinase N-associated protein, A-kinase anchoring protein 450, A-kinase anchor protein, 350kDa, protein kinase A anchoring protein 9, centrosome- and golgi-localized protein, AKAP450, MU-RMS-40.16A, A-kinase anchor protein 9, AKAP9-BRAF fusion protein |
| Gene Symbols | AKAP9 |
| Host Species | Rabbit |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.