Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ AKIP1 Monoclonal Antibody (OTI2E4)
GREENER_CHOICE

Catalog No. PIMA526998
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIMA526998 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIMA526998 Supplier Invitrogen™ Supplier No. MA526998
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

AKIP1 Monoclonal Antibody for IHC (P)

Enhances NF-kappa-B transcriptional activity by regulating the nuclear localization of the NF-kappa-B subunit RELA and promoting the phosphorylation of RELA by PRKACA. Regulates the effect of the cAMP-dependent protein kinase signaling pathway on the NF-kappa-B activation cascade.
TRUSTED_SUSTAINABILITY

Specifications

Antigen AKIP1
Applications Immunohistochemistry (Paraffin)
Classification Monoclonal
Clone OTI2E4
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 1% BSA, 50% glycerol and 0.02% sodium azide; pH 7.3
Gene Akip1
Gene Accession No. Q9NQ31
Gene Alias A kinase (PRKA) interacting protein 1; A-kinase interacting protein 1; A-kinase-interacting protein 1; Akip1; BCA3; breast cancer associated 3; breast cancer associated gene 3; breast cancer-associated gene 3 protein; C11orf17; D7H11orf17; D930014E17Rik; ICRFP703B1614Q5.6; ICRFP703N2430Q5.6; koyt binding protein 1; koyt binding protein 2; koyt binding protein 3; ORF27; PKA-interacting protein; Proline-rich protein BCA3; protein kinase A-interacting protein 1; RGD1306959
Gene Symbols Akip1
Host Species Mouse
Immunogen Full length human recombinant protein of AKIP1 produced in E.coli.
Purification Method Affinity Chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 56672
Target Species Human
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.