Learn More
Invitrogen™ AKIP1 Polyclonal Antibody

Description
Immunogen sequence: VDRRSLQRSAR LALEVLERAK RRAVDWHALE RPKGCMGVLA REAPHLEKQP AAGPQRVLPG E Highest antigen sequence identity to the following orthologs - mouse 63%, rat 63%.
Specifications
Specifications
| Antigen | AKIP1 |
| Applications | Immunocytochemistry |
| Classification | Polyclonal |
| Concentration | 0.05 mg/mL |
| Conjugate | Unconjugated |
| Formulation | PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 |
| Gene | Akip1 |
| Gene Accession No. | Q9NQ31 |
| Gene Alias | A kinase (PRKA) interacting protein 1; A-kinase interacting protein 1; A-kinase-interacting protein 1; Akip1; BCA3; breast cancer associated 3; breast cancer associated gene 3; breast cancer-associated gene 3 protein; C11orf17; D7H11orf17; D930014E17Rik; ICRFP703B1614Q5.6; ICRFP703N2430Q5.6; koyt binding protein 1; koyt binding protein 2; koyt binding protein 3; ORF27; PKA-interacting protein; Proline-rich protein BCA3; protein kinase A-interacting protein 1; RGD1306959 |
| Gene Symbols | Akip1 |
| Show More |
Safety and Handling
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.