Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
AKIRIN1 Antibody, Alexa Fluor™ 647, Novus Biologicals™

Rabbit Polyclonal Antibody
Supplier: Novus Biologicals NBP281699AF647
This item is not returnable.
View return policy
Description
AKIRIN1 Polyclonal antibody specifically detects AKIRIN1 in Human, Mouse, Rat samples. It is validated for Western Blot, ELISASpecifications
AKIRIN1 | |
Polyclonal | |
50mM Sodium Borate | |
Rabbit | |
Peptide affinity purified | |
RUO | |
Primary | |
Human, Mouse, Rat | |
Purified |
Western Blot, ELISA | |
Alexa Fluor 647 | |
AKIRIN1 akirin 1, C1orf108, RP11-781D11.2, STRF2 | |
AKIRIN1 antibody was raised against a 16 amino acid synthetic peptide near the amino terminus of the human AKIRIN1. The immunogen is located within the first 50 amino acids of AKIRIN1. Amino Acid Squence: LPGPTPGLRPPDAEPPP | |
0.1 mL | |
Innate Immunity | |
79647 | |
Store at 4°C in the dark. | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction