Learn More
Invitrogen™ Akirin2 Polyclonal Antibody

Description
A suggested positive control is human brain tissue lysate. PA5-20608 can be used with blocking peptide PEP-0728.
Specifications
Specifications
| Antigen | Akirin2 |
| Applications | Western Blot |
| Classification | Polyclonal |
| Concentration | 1 mg/mL |
| Conjugate | Unconjugated |
| Formulation | PBS with 0.02% sodium azide |
| Gene | AKIRIN2 |
| Gene Accession No. | B1AXD8, Q25C79, Q53H80 |
| Gene Alias | 14-3-3 beta; 2700059D21Rik; AA114675; AA522011; akirin 2; Akirin2; Akirin-2; akirin2 {ECO:0000250; AU019887; C6orf166; dJ486L4.2; EMBL:BAE81786.1}; FBI1; Fourteen-three-three beta interactant 1; fourteen-three-three beta interactant 1 {ECO:0000312; fourteen-three-three beta interacting-protein 1; RGD1307791; UniProtKB:Q53H80} |
| Gene Symbols | AKIRIN2 |
| Show More |
Safety and Handling
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.