Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

AKIRIN2 Rabbit anti-Human, Polyclonal, Bioss
SDP

Catalog No. 501988805
Encompass_Preferred
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-198-8805 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 50-198-8805 Supplier Bioss Supplier No. BS9082R
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

The highly conserved, nuclear-localized Akirin1 and Akirin2 proteins critically regulate the transcription of NF-kappa-B-dependent genes and are required for defense against Gram-negative bacteria in the immune deficiency and NF-kappa-B pathways. Akirin1 is dispensable in the mouse, and neither knockout mice nor cells derived from them have obvious distinctive phenotypes. In contrast, Akirin2 is required for development in the mouse and knockout of both Akirin homologs in mice show that Akirin2 is required downstream of toll-like receptor (TLR), TNF-a and IL-1-b signaling, and for the production of IL-6. Akirin2 is functionally closer to the single gene in Drosophila, as the homozygous null D. melanogaster Akirin mutants show a similar, mid-to-early embryonic death.

Specifications

Antigen AKIRIN2
Applications Immunofluorescence, Immunohistochemistry (Paraffin), Western Blot
Classification Polyclonal
Concentration 1 μg/mL
Conjugate Unconjugated
Formulation PBS with 1% BSA, 50% glycerol and 0.09% sodium azide; pH
Gene AKIRIN2
Gene Accession No. Q53H80
Gene Alias 14-3-3 beta; 2700059D21Rik; AA114675; AA522011; akirin 2; AKIRIN2; Akirin-2; akirin2 {ECO:0000250; AU019887; C6orf166; dJ486L4.2; EMBL:BAE81786.1}; FBI1; Fourteen-three-three beta interactant 1; fourteen-three-three beta interactant 1 {ECO:0000312; Fourteen-three-three beta interacting-protein 1; RGD1307791; UniProtKB:Q53H80}
Gene Symbols AKIRIN2
Host Species Rabbit
Immunogen KLH conjugated synthetic peptide derived from human AKIRIN2.
Purification Method Protein A
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 55122
Target Species Human
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.