Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ AKNA Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA566652
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA566652 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA566652 Supplier Invitrogen™ Supplier No. PA566652
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: RLSHISTAGTL AQPFAASVPR DGASYPKARG SLIPRRATEP STPRSQAQRY LSSPSGPLRQ RAPNFSLERT LAAEMAVPGS EFEGHKRISE QPL Highest antigen sequence identity to the following orthologs - mouse 54%, rat 51%.

AKNA is a transcription factor that specifically activates the expression of the CD40 receptor and its ligand CD40L/CD154, two cell surface molecules on lymphocytes that are critical for antigen-dependent-B-cell development. It binds to A/T-rich promoters.
TRUSTED_SUSTAINABILITY

Specifications

Antigen AKNA
Applications Immunocytochemistry
Classification Polyclonal
Concentration 0.3 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene AKNA
Gene Accession No. Q7Z591
Gene Alias AKNA; AKNA transcript F2; AT-hook transcription factor; AT-hook transcription factor AKNA; AT-hook-containing transcription factor; KIAA1968; Microtubule organization protein AKNA
Gene Symbols AKNA
Host Species Rabbit
Immunogen Recombinant Human AKNA. Recombinant protein control fragment (Product #RP-105295).
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 80709
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.