Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ AKR1A1 Polyclonal Antibody

Goat Polyclonal Antibody
Supplier: Invitrogen™ PA518073
Description
This antibody is predicted to react with bovine, canine, mouse, porcine and rat based on sequence homology. This antibody is tested in Peptide ELISA: antibody detection limit dilution 16,000.
This gene encodes a member of the aldo/keto reductase superfamily, which consists of more than 40 known enzymes and proteins. This member, also known as aldehyde reductase, is involved in the reduction of biogenic and xenobiotic aldehydes and is present in virtually every tissue. Multiple alternatively spliced transcript variants of this gene exist, all encoding the same protein.
Specifications
| AKR1A1 | |
| Polyclonal | |
| Unconjugated | |
| AKR1A1 | |
| 2610201A18Rik; 3-DG-reducing enzyme; Akr1a1; AKR1A1 protein; Akr1a4; alcohol dehydrogenase; alcohol dehydrogenase [NADP(+)]; Aldehyde reductase; aldo-keto reductase family 1 member A1; aldo-keto reductase family 1, member A1 (aldehyde reductase); aldo-keto reductase family 1, member A4 (aldehyde reductase); ALDR1; ALR; ARM; DD3; dihydrodiol dehydrogenase 3; epididymis secretory protein Li 6; epididymis secretory sperm binding protein Li 165mP; Glucuronate reductase; Glucuronolactone reductase; HEL-S-165mP; HEL-S-6; MGC12529; MGC1380; OTTHUMP00000009240 | |
| Goat | |
| Ammonium Sulfate Precipitation | |
| RUO | |
| 10327 | |
| -20°C, Avoid Freeze/Thaw Cycles | |
| Liquid |
| Immunohistochemistry (Paraffin), Western Blot | |
| 0.5 mg/mL | |
| TBS with 0.5% BSA and 0.02% sodium azide; pH 7.3 | |
| P14550 | |
| AKR1A1 | |
| Synthetic peptide sequence (DAGHPLYPFNDPY) corresponding to the C-terminus amino acids of AKR1A1 (aa 313-325). | |
| 100 μg | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction