Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

AKR1B1 Mouse anti-Human, Unconjugated, Clone: CPTC-AKR1B1-2, Abnova™

Catalog No. 89337471 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-337-471 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-337-471 Supplier Abnova Corporation Supplier No. MAB20976
Add to Cart
Add to Cart

Mouse Monoclonal Antibody.

AKR1B1 Monoclonal antibody specifically detects AKR1B1 in Human samples. It is validated for WB, IHC-P.

Specifications

Antigen AKR1B1
Applications Western Blot, Immunohistochemistry (Paraffin)
Classification Monoclonal
Clone CPTC-AKR1B1-2
Conjugate Unconjugated
Description aldo-keto reductase family 1, member B1 (aldose reductase)
Gene AKR1B1
Gene Alias ADR, ALDR1, ALR2, AR, MGC1804
Host Species Mouse
Immunogen Recombinant protein corresponding to full length human AKR1B1
Quantity 100 μg
Primary or Secondary Primary
Gene ID (Entrez) 231
Target Species Human
Content And Storage Store at -20 to -80°C. Aliquot to avoid repeated freezing and thawing.
Form Liquid
Isotype IgG1 κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.