Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ AKR1B1 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA584125
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA584125 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA584125 Supplier Invitrogen™ Supplier No. PA584125
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: VTPERIAENF KVFDFELSSQ DMTTLLSYNR NWRVCALLSC TSHKDY Highest antigen sequence indentity to the following orthologs: Mouse - 83%, Rat - 85%.

Catalyzes the NADPH-dependent reduction of a wide variety of carbonyl-containing compounds to their corresponding alcohols with a broad range of catalytic efficiencies.
TRUSTED_SUSTAINABILITY

Specifications

Antigen AKR1B1
Applications Immunohistochemistry (Paraffin), Western Blot
Classification Polyclonal
Concentration 0.05 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene AKR1B1
Gene Accession No. P15121
Gene Alias ADR; Ahr1; Ahr-1; Akr1b1; Akr1b3; Akr1b4; Aldehyde reductase; aldehyde reductase 1; Aldehyde reductase 1 (low Km aldose reductase) (5.8 kb PstI fragment, probably the functional gene); aldo-keto reductase family 1; aldo-keto reductase family 1 member B; aldo-keto reductase family 1 member B1; aldo-keto reductase family 1 member B1; aldose reductase; Aldo-keto reductase family 1 member B3; aldo-keto reductase family 1, member B1 (aldose reductase); aldo-keto reductase family 1, member B3 (aldose reductase); aldo-keto reductase family 1, member B4 (aldose reductase); Aldor1; Aldose reductase; ALDR1; ALDRED; Alr; Alr2; ALR-P-I; AR; Lii5-2 CTCL tumor antigen; low Km aldose reductase; member B1 (aldose reductase); RATALDRED
Gene Symbols AKR1B1
Host Species Rabbit
Immunogen Recombinant protein corresponding to Human AKR1B1. Recombinant protein control fragment (Product #RP-104952).
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 231
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.