Learn More
AKR1B1 Rabbit anti-Human, Polyclonal, Bethyl Laboratories
Rabbit Polyclonal Antibody
Supplier: Bethyl Laboratories, Inc A305312A

Description
The recommended shelf life for this product is 1 year from date of receipt.
Catalyzes the NADPH-dependent reduction of a wide variety of carbonyl-containing compounds to their corresponding alcohols with a broad range of catalytic efficiencies.Specifications
AKR1B1 | |
Polyclonal | |
Unconjugated | |
AKR1B1 | |
Aldose reductase; AR; Aldehyde reductase; AKR1B1 | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
231 | |
4° C | |
Liquid |
Western Blot | |
1 mg/ml | |
phosphate, tris citrate with 0.09% sodium azide; pH 7-8 | |
P15121 | |
AKR1B1 | |
Between 100 and 150 | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.