Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
AKR1B1 Rabbit anti-Human, Clone: 101, Novus Biologicals™

Rabbit Monoclonal Antibody
$342.00 - $524.00
Specifications
Antigen | AKR1B1 |
---|---|
Clone | 101 |
Dilution | Flow Cytometry 1:25-1:100 |
Applications | Flow Cytometry |
Classification | Monoclonal |
Description
AKR1B1 Monoclonal specifically detects AKR1B1 in Human samples. It is validated for Flow Cytometry.Specifications
AKR1B1 | |
Flow Cytometry 1:25-1:100 | |
Monoclonal | |
Purified | |
RUO | |
Human | |
ADR, Aldehyde reductase, Aldo-keto reductase family 1 member B1, aldo-keto reductase family 1, member B1 (aldose reductase), ALDR1aldose reductase, ALR2, ARaldehyde reductase 1, EC 1.1.1, EC 1.1.1.21, Lii5-2 CTCL tumor antigen, low Km aldose reductase, MGC1804 | |
Recombinant Human AKR1B1 Protein | |
Primary | |
Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles. |
101 | |
Flow Cytometry | |
Unconjugated | |
Rabbit | |
Stem Cell Markers | |
0.2 um filtered solution in PBS | |
231 | |
IgG | |
Protein A purified |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title