Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ AKR1B10 Polyclonal Antibody
Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA597444
Description
AKR1B10 Polyclonal Antibody for Western Blot, IHC (P), ELISA
AKR1B10, a 316aa protein, catalyzes efficient reduction ofaliphatic and aromatic aldehydes. It the first cytosolic NADP (H)-dependent retinal reductase described in humans and is characterized by the presence of a novel NADP binding motif. Due to its ability to convert glucose to sorbitol, AKR1B10 may have a role in development of secondary diabetic complications.It is abundantly expressed in adrenal gland, small intestine and colon, with lower levels in liver, thymus, prostate, testis, and skeletal muscle and its levels are up regulated in certain cancers such as hepatocellular carcinoma.
Specifications
AKR1B10 | |
Polyclonal | |
Unconjugated | |
Akr1b10 | |
2310005E10Rik; AKR1B10; AKR1B11; AKR1B12; Akr1b16; aldo-keto reductase family 1 member B10; aldo-keto reductase family 1, member B10; aldo-keto reductase family 1, member B10 (aldose reductase); aldo-keto reductase family 1, member B11 (aldose reductase-like); Aldose reductase-like; aldose reductase-like 1; aldose reductase-like peptide; aldose reductase-related protein; ALDRLn; ARL1; ARL-1; ARP; hARP; HIS; HSI; MGC14103; SI reductase; small intestine reductase | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
57016 | |
-20°C or -80°C if preferred | |
Liquid |
ELISA, Immunohistochemistry (Paraffin), Western Blot | |
3.88 mg/mL | |
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
O60218 | |
Akr1b10 | |
Recombinant Human Aldo-keto reductase family 1 member B10 protein (1-316AA). | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction