Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ AKR1B8 Polyclonal Antibody

Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA5114302
Description
Immunogen sequence: TNQVECHPYL TQEKLIQYCH SKGISVTAYS PLGSPDRPSA KPEDPSLLED For short term use, store at 2-8°C up to 1 week. For long term storage, store at -20°C in small aliquots to prevent freeze-thaw cycles.
The function of this protein remains unknown.
Specifications
| AKR1B8 | |
| Polyclonal | |
| Unconjugated | |
| AKR1B8 | |
| Akr1b8; Aldehyde reductase; aldo-keto reductase family 1, member B8; aldose reductase-related protein 2; AR; Fgfrp; Fgrp; fibroblast growth factor regulated protein; fibroblast growth factor-regulated protein; FR-1; Protein FR-1 | |
| Rabbit | |
| Affinity chromatography | |
| RUO | |
| 14187 | |
| -20°C, Avoid Freeze/Thaw Cycles | |
| Liquid |
| Western Blot | |
| 0.5 mg/mL | |
| PBS with 2% sucrose and 0.09% sodium azide | |
| P45377 | |
| AKR1B8 | |
| Synthetic peptide directed towards the middle terminal region of mouse AKR1B8 (aa 182-231). | |
| 100 μL | |
| Primary | |
| Mouse | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction