Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ AKR1C1 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA5120192
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA5120192 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. PIPA5120192 Supplier Invitrogen™ Supplier No. PA5120192
Add to Cart
Add to Cart
This item is not returnable. View return policy

Rabbit Polyclonal Antibody

Positive test controls include: HeLa, 22Rv1, U-87MG, Mouse liver. The target is usually found in the following locations: Cytoplasm. Immunogen sequence: MDSKYQCVKL NDGHFMPVLG FGTYAPAEVP KSKALEATKL AIEAGFRHID SAHLYNNEEQ VGLAIRSKIA DGSVKREDIF YTSKLWCNSH RPELVRPALE RSLKNLQLDY VDLYLIHFPV SVKPGEEVIP KDENGKILFD TVDLCATWEA VEKCKDAGLA KSIGVSNFNR RQLEMILNKP GLKYKPVCNQ VECHPYFNQR KLLDFCKSKD IVLVAYSALG SHREEPWVDP NSPVLLEDPV LCALAKKHKR TPALIALRYQ LQRGVVVLAK SYNEQRIRQN VQVFEFQLTS EEMKAIDGLN RNVRYLTLDI FAGPPNYPFS DEY.

This gene encodes a member of the aldo/keto reductase superfamily, which consists of more than 40 known enzymes and proteins. These enzymes catalyze the conversion of aldehydes and ketones to their corresponding alcohols by utilizing NADH and/or NADPH as cofactors. The enzymes display overlapping but distinct substrate specificity. This enzyme catalyzes the reaction of progesterone to the inactive form 20-alpha-hydroxy-progesterone. This gene shares high sequence identity with three other gene members and is clustered with those three genes at chromosome 10p15-p14.
TRUSTED_SUSTAINABILITY

Specifications

Antigen AKR1C1
Applications ELISA, Western Blot
Classification Polyclonal
Concentration 0.22 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.01% thimerosal; pH 7.3
Gene AKR1C1
Gene Accession No. P70694, Q04828
Gene Alias 17-beta-HSD 5; 17HSD type 5; 20 alpha-hydroxysteroid dehydrogenase; 20-alpha (3-alpha)-hydroxysteroid dehydrogenase); 20-ALPHA-HSD; 20-alpha-hydroxysteroid dehydrogenase; 2-ALPHA-HSD; 3alpha-HSD; Akr1c1; Akr1c6; aldo-keto reductase C; aldo-keto reductase family 1 member C1; aldo-keto reductase family 1, member C1; aldo-keto reductase family 1, member C1 (aldehyde reductase); aldo-keto reductase family 1, member C1 (dihydrodiol dehydrogenase 1; aldo-keto reductase family 1, member C1 (dihydrodiol dehydrogenase 1; 20-alpha (3-alpha)-hydroxysteroid dehydrogenase); aldo-keto reductase family 1, member C6; C9; chlordecone reductase homolog HAKRC; DD1; DD1/DD2; DDH; DDH1; dihydrodiol dehydrogenase 1; dihydrodiol dehydrogenase 1/2; dihydrodiol dehydrogenase 1; 20-alpha (3-alpha)-hydroxysteroid dehydrogenase; dihydrodiol dehydrogenase isoform DD1; estradiol 17 beta-dehydrogenase 5; estradiol 17-beta-dehydrogenase (A-specific); H-37; HAKRC; HBAB; hepatic dihydrodiol dehydrogenase; high-affinity hepatic bile acid-binding protein; Hsd17b5; hydroxysteroid 17-beta dehydrogenase 5; indanol dehydrogenase; MBAB; RP11-69D4.1; trans-1,2-dihydrobenzene-1,2-diol dehydrogenase; type 5 17beta-hydroxysteroid dehydrogenase; type II 3-alpha-hydroxysteroid dehydrogenase
Gene Symbols AKR1C1, Akr1c6
Host Species Rabbit
Immunogen Recombinant fusion protein containing a sequence corresponding to amino acids 1-323 of human AKR1C1 (NP_001344.2).
Purification Method Affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 1645, 83702
Target Species Human, Mouse
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.