Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ AKR1C2 Polyclonal Antibody
Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA5101414
Description
Antibody detects endogenous levels of total AKR1C2.
DD1 is also designated AKR1C1, DDH or DDH1, while DD2 also can be designated AKR1C2, dDD, BABP or DDH2. AKR1C3 and 3 alpha-HSD are alternate designations for human DD3 (which is referred to as AKR1C18 in rodents), while DD4 also can be called AKR1C4, CD, CHDR or AKR1C6 (in rodents). DD1 and DD2 are 20 alpha-HSDs, whereas DD3 and DD4 are the 3 alpha- HSDs. The multiple human cytosolic dihydrodiol dehydrogenases are involved in the metabolism of xenobiotics, such as polycyclic aromatic hydrocarbons, pesticides and steroid hormones, and are responsible for the reduction of ketone-containing drugs by using NADH or NADPH as a cofactor. The 20 alpha- HSD catalyzes the reaction of Progesterone to the inactive form 20 alpha-hydroxyprogesterone.
Specifications
AKR1C2 | |
Polyclonal | |
Unconjugated | |
AKR1C2 | |
17-alpha-HSD; 17-alpha-hydroxysteroid dehydrogenase; 3 alpha-HSD3; 3(17)alpha-hydroxysteroid dehydrogenase; 3(or 17)-alpha-hydroxysteroid dehydrogenase; 3-alpha-HSD3; 3-alpha-hydroxysteroid dehydrogenase; 9430025F20Rik; AI315367; AKR1C2; Akr1c21; AKR1C-pseudo; Aldo-keto reductase family 1 member C2; aldo-keto reductase family 1 member C21; aldo-keto reductase family 1, member C2; aldo-keto reductase family 1, member C21; BABP; Chlordecone reductase homolog HAKRD; DD; DD/BABP; DD1; DD2; DD-2; DD3; DDH2; Dihydrodiol dehydrogenase 2; dihydrodiol dehydrogenase 2; bile acid binding protein; 3-alpha hydroxysteroid dehydrogenase, type III; dihydrodiol dehydrogenase type 1; Dihydrodiol dehydrogenase type 3; Dihydrodiol dehydrogenase/bile acid-binding protein; HAKRD; HBAB; HSD3-3 alpha; HSD3-alpha3; MCDR2; pseudo-chlordecone reductase; SRXY8; TDD; testicular 17,20-desmolase deficiency; trans-1,2-dihydrobenzene-1,2-diol dehydrogenase; type II dihydrodiol dehydrogenase; Type III 3-alpha-hydroxysteroid dehydrogenase | |
Rabbit | |
Affinity chromatography | |
RUO | |
1646 | |
-20°C | |
Liquid |
Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry | |
1 mg/mL | |
PBS with 50% glycerol and 0.02% sodium azide; pH 7.4 | |
P52895 | |
AKR1C2 | |
A synthesized peptide derived from human AKR1C2(Accession P52895), corresponding to amino acid residues E28-D78. | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Safety and Handling
WARNING: Cancer - www.P65Warnings.ca.gov
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction