Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

AKR1C3 Rabbit anti-Human, Polyclonal, Proteintech
SDP

Catalog No. 501726958 Shop All Proteintech Group Inc Products
Encompass_Preferred
Change view
Click to view available options
Quantity:
150 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-172-6958 150 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 50-172-6958 Supplier Proteintech Group Inc Supplier No. 111941AP150UL
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

DD1 is also designated AKR1C1, DDH or DDH1 while DD2 also can be designated AKR1C2, dDD, BABP or DDH2. AKR1C3 and 3alpha-HSD are alternate designations for DD3, while DD4 also can be called AKR1C4, CD or CHDR. DD3 is a unique enzyme that can specifically catalyze the dehydrogenation of trans-benzenedihydrodiol and trans-naphthalenedihydrodiol.

Specifications

Antigen AKR1C3
Applications Immunohistochemistry (Paraffin), Immunoprecipitation, Western Blot
Classification Polyclonal
Concentration 0.33 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.1% sodium azide; pH 7.3
Gene AKR1C3
Gene Accession No. P42330
Gene Alias 17 beta HSD 5, 3 alpha HSD type 2, 3 alpha HSD type II, brain, AKR1C3, DD 3, DD3, DDX, Dihydrodiol dehydrogenase 3, HA1753, HAKRB, HAKRe, hluPGFS, HSD17B5, Indanol dehydrogenase, KIAA0119, PGFS, Prostaglandin F synthase
Gene Symbols AKR1C3
Host Species Rabbit
Immunogen AKR1C3 Fusion Protein Ag1674
Purification Method Antigen Affinity Chromatography
Quantity 150 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 8644
Target Species Human
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.