Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ AKR1C4 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA5103590
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA5103590 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA5103590 Supplier Invitrogen™ Supplier No. PA5103590
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Antibody detects endogenous levels of total AKR1C4.

This gene encodes a member of the aldo/keto reductase superfamily, which consists of more than 40 known enzymes and proteins. These enzymes catalyze the conversion of aldehydes and ketones to their corresponding alcohols by utilizing NADH and/or NADPH as cofactors. The enzymes display overlapping but distinct substrate specificity. This enzyme catalyzes the bioreduction of chlordecone, a toxic organochlorine pesticide, to chlordecone alcohol in liver. This gene shares high sequence identity with three other gene members and is clustered with those three genes at chromosome 10p15-p14.
TRUSTED_SUSTAINABILITY

Specifications

Antigen AKR1C4
Applications Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry
Classification Polyclonal
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.02% sodium azide; pH 7.4
Gene AKR1C4
Gene Accession No. P17516
Gene Alias 3-alpha-HSD; 3-alpha-HSD1; 3-alpha-hydroxysteroid dehydrogenase type I; AKR1C4; Aldo-keto reductase family 1 member C4; aldo-keto reductase family 1, member C4; C11; CDR; CHDR; Chlordecone reductase; chlordecone reductase; 3-alpha hydroxysteroid dehydrogenase, type I; dihydrodiol dehydrogenase 4; DD4; DD-4; Dihydrodiol dehydrogenase 4; dihydrodiol dehydrogenase isozyme DD4; HAKRA; HGNC:387; MGC22581; type I 3-alpha-hydroxysteroid dehydrogenase
Gene Symbols AKR1C4
Host Species Rabbit
Immunogen A synthesized peptide derived from human AKR1C4(Accession P17516), corresponding to amino acid residues M1-N32.
Purification Method Affinity Chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 1109
Target Species Human, Rat
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.