Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ AKR1D1 Polyclonal Antibody
Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA563384
Description
Immunogen sequence: MDLSAASHRI PLSDGNSIPI IGLGTYSEPK STPKGACATS VKVAIDTG Highest antigen sequence identity to the following orthologs: Mouse - 63%, Rat - 65%.
The enzyme encoded by this gene is responsible for the catalysis of the 5-beta-reduction of bile acid intermediates and steroid hormones carrying a delta(4)-3-one structure. Deficiency of this enzyme may contribute to hepatic dysfunction.
Specifications
AKR1D1 | |
Polyclonal | |
Unconjugated | |
AKR1D1 | |
3o5bred; 3-oxo-5-beta-steroid 4-dehydrogenase; Akr1d1; Aldo-keto reductase family 1 member D1; aldo-keto reductase family 1, member D1; aldo-keto reductase family 1, member D1 (delta 4-3-ketosteroid-5-beta-reductase); CBAS2; delta 4-3-ketosteroid-5-beta-reductase; delta(4)-3-ketosteroid 5-beta-reductase; Delta(4)-3-oxosteroid 5-beta-reductase; SRD5B1; steroid-5-beta-reductase, beta polypeptide 1 (3-oxo-5 beta-steroid delta 4-dehydrogenase beta 1) | |
Rabbit | |
Antigen Affinity Chromatography | |
RUO | |
6718 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Immunohistochemistry (Paraffin), Western Blot | |
0.05 mg/mL | |
PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
P51857 | |
AKR1D1 | |
Recombinant protein corresponding to Human AKR1D1. Recombinant protein control fragment (Product #RP-100116). | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction