Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ AKR1E2 Polyclonal Antibody
Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA558123
Description
Immunogen sequence: MGFKPPHPEW IMSCSELSFC LSHPRVQDLP LDESNMVIPS DTDFLDTWEA MEDLVITGLV KNIG Highest antigen sequence identity to the following orthologs: Mouse - 44%, Rat - 48%.
The protein encoded by this gene is a member of the aldo-keto reductase superfamily. Members in this family are characterized by their structure (evolutionarily highly conserved TIM barrel) and function (NAD(P)H-dependent oxido-reduction of carbonyl groups). Transcripts of this gene have been reported in specimens of human testis. Alternative splicing results in multiple transcript variants.
Specifications
AKR1E2 | |
Polyclonal | |
Unconjugated | |
AKR1E2 | |
1,5-anhydro-D-fructose reductase; AF reductase; Akr1cl2; Akr1e1; Akr1e2; AKRDC1; Aldo-keto reductase family 1 member C-like protein 2; aldo-keto reductase family 1 member E2; aldo-keto reductase family 1, member C-like 2; aldo-keto reductase family 1, member E1; aldo-keto reductase family 1, member E2; aldo-keto reductase loopADR; htAKR; hTSP; LoopADR; Testis aldo-keto reductase; Testis-specific protein | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
83592 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Immunohistochemistry (Paraffin), Western Blot | |
0.3 mg/mL | |
PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
Q96JD6 | |
AKR1E2 | |
Recombinant protein corresponding to Human AKR1E2. Recombinant protein control fragment (Product #RP-97128). | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction