Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ AKR1E2 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA583392
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA583392 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA583392 Supplier Invitrogen™ Supplier No. PA583392
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: LTQKNLISFC QSRDVSVTAY RPLGGSCEGV DLIDNPVIKR IAKEHGKSPA QI Highest antigen sequence indentity to the following orthologs: Mouse - 69%, Rat - 71%.

The protein encoded by this gene is a member of the aldo-keto reductase superfamily. Members in this family are characterized by their structure (evolutionarily highly conserved TIM barrel) and function (NAD(P)H-dependent oxido-reduction of carbonyl groups). Transcripts of this gene have been reported in specimens of human testis. Alternative splicing results in multiple transcript variants.
TRUSTED_SUSTAINABILITY

Specifications

Antigen AKR1E2
Applications Immunocytochemistry
Classification Polyclonal
Concentration 0.3 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene AKR1E2
Gene Accession No. Q96JD6
Gene Alias 1,5-anhydro-D-fructose reductase; AF reductase; Akr1cl2; Akr1e1; Akr1e2; AKRDC1; Aldo-keto reductase family 1 member C-like protein 2; aldo-keto reductase family 1 member E2; aldo-keto reductase family 1, member C-like 2; aldo-keto reductase family 1, member E1; aldo-keto reductase family 1, member E2; aldo-keto reductase loopADR; htAKR; hTSP; LoopADR; Testis aldo-keto reductase; Testis-specific protein
Gene Symbols AKR1E2
Host Species Rabbit
Immunogen Recombinant protein corresponding to Human AKR1E2. Recombinant protein control fragment (Product #RP-104680).
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 83592
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.